accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117772-70-0

H2OH2OCH3(R)OO(R)H3C(S)O(R)(R)H3COH3C(S)OH(S)CH3O(S)H3C(R)O(S)O(R)CH3(S)(R)HONCH3H3C(R)CH3HO(R)CH3NH3C(R)CH3(R)HO(S)H3COH

CAS RN 117772-70-0

IUPAC Name: (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-2-ethyl-3,4,10-trihydroxy-13-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one dihydrate
Synonyms: tromix azitromax azitroxil aciphar azitrix azitral azitrom azitrox acitrocin sumamed
Molecular Weight (g/mol): 785.03
Molecular Formula: C38H76N2O14
InChi Key: SRMPHJKQVUDLQE-KUJJYQHYSA-N
SMILES: O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products