CAS RN 118290-26-9
CAS RN 118290-26-9
IUPAC Name:
N-(2-{2-[(dipropylamino)methyl]piperidin-1-yl}ethyl)-10-oxo-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-2-carboxamide
Synonyms:
N-(2-{2-[(dipropylamino)methyl]piperidin-1-yl}ethyl)-10-oxo-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-2-carboxamide
Molecular Weight (g/mol):
478.64
Molecular Formula:
C27H38N6O2
InChi Key:
MZDYABXXPZNUCT-UHFFFAOYNA-N
SMILES:
CCCN(CCC)CC1CCCCN1CCNC(=O)N1C2=CC=CC=C2C(=O)NC2=C1N=CC=C2
1
–
1
of
1
results
1
AF-DX 384, Tocris Bioscience™
CAS: 118290-26-9 Molecular Formula: C27H38N6O2 Molecular Weight (g/mol): 478.641 InChI Key: MZDYABXXPZNUCT-UHFFFAOYSA-N Synonym: af-dx 384,afdx384,af-dx-384,3h af dx-384,3h af-dx 384,n-2-2-dipropylamino methyl piperidin-1-yl ethyl-6-oxo-5h-benzo e pyrido 3,2-b 1,4 diazepine-11 6h-carboxamide,n-2-2-dipropylamino methyl-1-piperidinyl ethyl-5,6-dihydro-6-oxo-11h-pyrido 2,3-b 1,4 benzodiazepine-11-carboxamide,afdx-384,n-2-2-dipropylamino methyl piperidin-1-yl ethyl-6-oxo-5h-pyrido 2,3-b 1,4 benzodiazepine-11-carboxamide,d06pvq PubChem CID: 119357 IUPAC Name: N-[2-[2-[(dipropylamino)methyl]piperidin-1-yl]ethyl]-6-oxo-5H-pyrido[2,3-b][1,4]benzodiazepine-11-carboxamide SMILES: CCCN(CCC)CC1CCCCN1CCNC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4