CAS RN 118362-03-1
CAS RN 118362-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific™ TheraPure™ rU Phosphoramidite, 20-400 finish bottle
rU CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) for use in oligonucleotide synthesis.
Promotions are available