accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1185301-51-2

NHOClHCl

CAS RN 1185301-51-2

IUPAC Name: 3-(3-chlorophenoxy)piperidine hydrochloride
Synonyms: 3-(3-chlorophenoxy)piperidine hydrochloride
Molecular Weight (g/mol): 248.15
Molecular Formula: C11H15Cl2NO
InChi Key: CNQOQASAMXQVLU-UHFFFAOYNA-N
SMILES: Cl.ClC1=CC=CC(OC2CCCNC2)=C1
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products