man

Test

CAS RN 1187460-81-6

H3CNH3CHONOHNNNOH2NFClOH

CAS RN 1187460-81-6

IUPAC Name: 2-[3,4-bis(hydroxymethyl)-3,4-dimethylpyrrolidin-1-yl]-N-[3-carbamoyl-1-(4-fluorophenyl)-1H,4H,5H-benzo[g]indazol-8-yl]-5-chloropyridine-4-carboxamide
Synonyms: 2-[3,4-bis(hydroxymethyl)-3,4-dimethylpyrrolidin-1-yl]-N-[3-carbamoyl-1-(4-fluorophenyl)-4H,5H-benzo[g]indazol-8-yl]-5-chloropyridine-4-carboxamide
Molecular Weight (g/mol): 619.09
Molecular Formula: C32H32ClFN6O4
InChi Key: JUOWWGNRWRLBSV-UHFFFAOYNA-N
SMILES: CC1(CO)CN(CC1(C)CO)C1=NC=C(Cl)C(=C1)C(=O)NC1=CC2=C(CCC3=C2N(N=C3C(N)=O)C2=CC=C(F)C=C2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results