man

Test

CAS RN 118799-24-9

CH3NCH3H3N

CAS RN 118799-24-9

IUPAC Name: (1,5-dimethyl-1H-pyrrol-2-yl)methanaminium
Synonyms: (1,5-dimethylpyrrol-2-yl)methanaminium
Molecular Weight (g/mol): 125.19
Molecular Formula: C7H13N2
InChi Key: KMUHAJOXPNPERW-UHFFFAOYSA-O
SMILES: CN1C(C)=CC=C1C[NH3+]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results