man

Test

CAS RN 1188-37-0

H3CONHOHOOHO

CAS RN 1188-37-0

IUPAC Name: 2-acetamidopentanedioic acid
Synonyms: acetylglutamate Ac-Glu-OH (D)-N-acetylglutamate N-acetylglutamate, (D)- N-ac-glu-OH 2-acetamido-L-glutaraldehydate N-acetylglutamate, (DL)-isomer N-acetylglutamate, (D)-isomer (D)-isomer N-acetylglutamate N-acetylglutamate, (DL)-
Molecular Weight (g/mol): 189.17
Molecular Formula: C7H11NO5
InChi Key: RFMMMVDNIPUKGG-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCC(O)=O)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)