accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1188365-70-9

NBrN

CAS RN 1188365-70-9

IUPAC Name: 6-bromoquinoline-5-carbonitrile
Synonyms: 6-bromoquinoline-5-carbonitrile
Molecular Weight (g/mol): 233.07
Molecular Formula: C10H5BrN2
InChi Key: DTWHKRFZYKUTHB-UHFFFAOYSA-N
SMILES: BrC1=C(C#N)C2=CC=CN=C2C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products