CAS RN 1194-64-5
CAS RN 1194-64-5
IUPAC Name:
2-chloro-6-methylbenzaldehyde
Synonyms:
2-chloro-6-methylbenzaldehyde
Molecular Weight (g/mol):
154.59
Molecular Formula:
C8H7ClO
InChi Key:
CCYFXIJPJFSTSU-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C(Cl)=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Chloro-6-methylbenzaldehyde, 98%
CAS: 1194-64-5 Molecular Formula: C8H7ClO Molecular Weight (g/mol): 154.593 MDL Number: MFCD01934409 InChI Key: CCYFXIJPJFSTSU-UHFFFAOYSA-N PubChem CID: 12467050 IUPAC Name: 2-chloro-6-methylbenzaldehyde SMILES: CC1=C(C(=CC=C1)Cl)C=O