man

Test

CAS RN 1195-81-9

CH3OOOH

CAS RN 1195-81-9

IUPAC Name: ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate
Synonyms: ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: PWMQFMMZBJUHID-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CO)CCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results