accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1197981-08-0

H3COSiOOFFFFFOH3COCH3

CAS RN 1197981-08-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 11-(triethoxysilyl)undecanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 11-(triethoxysilyl)undecanoate
Molecular Weight (g/mol): 514.61
Molecular Formula: C23H35F5O5Si
InChi Key: WQFFEEIHQJVCHR-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F)(OCC)OCC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products