CAS RN 1199523-24-4
CAS RN 1199523-24-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Cruzain-IN-1, MedChemExpress
MedChemExpress Cruzain-IN-1 is a covalent and reversible Cruzain inhibitor, with an IC50 of 10 nM.
