accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12003-96-2

AlTiTiTi

CAS RN 12003-96-2

IUPAC Name: aluminium(3+) trititanium
Synonyms: aluminium(3+) trititanium
Molecular Weight (g/mol): 170.58
Molecular Formula: AlTi3
InChi Key: JRDVYNLVMWVSFK-UHFFFAOYSA-N
SMILES: [Al+3].[Ti].[Ti].[Ti]
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products