accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12004-37-4

OOOOOAlAlSrSr

CAS RN 12004-37-4

IUPAC Name: dialuminium(3+) distrontium(2+) pentaoxidandiide
Synonyms: strontium aluminate
Molecular Weight (g/mol): 309.2
Molecular Formula: Al2O5Sr2
InChi Key: FNWBQFMGIFLWII-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Sr++].[Sr++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products