accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12006-85-8

BFeFe

CAS RN 12006-85-8

IUPAC Name: boron diiron
Synonyms: boron diiron
Molecular Weight (g/mol): 122.5
Molecular Formula: BFe2
InChi Key: FSDZRQFSRALZQJ-UHFFFAOYSA-N
SMILES: [B].[Fe].[Fe]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products