accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12007-00-0

BNi

CAS RN 12007-00-0

IUPAC Name: nickelioborane
Synonyms: nickelioborane
Molecular Weight (g/mol): 69.5
Molecular Formula: BNi
InChi Key: QDWJUBJKEHXSMT-UHFFFAOYSA-N
SMILES: B#[Ni]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products