accessibility menu, dialog, popup

UserName

Save up to 60% on cold storage essentials

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12017-01-5

OOOTiCo

CAS RN 12017-01-5

IUPAC Name: titanium(4+) λ²-cobalt(2+) trioxidandiide
Synonyms: cobalt titanium oxide
Molecular Weight (g/mol): 154.8
Molecular Formula: CoO3Ti
InChi Key: LFSBSHDDAGNCTM-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ti+4].[Co++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products