man

Test

CAS RN 120278-07-1

NNH2OO

CAS RN 120278-07-1

IUPAC Name: benzyl 4-aminopiperidine-1-carboxylate
Synonyms: benzyl 4-aminopiperidine-1-carboxylate
Molecular Weight (g/mol): 234.3
Molecular Formula: C13H18N2O2
InChi Key: YYIQGSYCCNQAGV-UHFFFAOYSA-N
SMILES: NC1CCN(CC1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results