man

Test

CAS RN 1204-60-0

O

CAS RN 1204-60-0

IUPAC Name: [1,1'-biphenyl]-3-carbaldehyde
Synonyms: [1,1'-biphenyl]-3-carbaldehyde
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: KFKSIUOALVIACE-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results