man

Test

CAS RN 1206593-30-7

CH3OBrFFF

CAS RN 1206593-30-7

IUPAC Name: 4-(bromomethyl)-1-ethoxy-2-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-1-ethoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 283.09
Molecular Formula: C10H10BrF3O
InChi Key: GAZKSECNCUGHQP-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(CBr)C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results