accessibility menu, dialog, popup

UserName

CAS RN 120686-18-2

CH3H3CCH3OO(S)NH3

CAS RN 120686-18-2

IUPAC Name: (1S)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Synonyms: (1S)-3-(tert-butoxy)-3-oxo-1-phenylpropan-1-aminium
Molecular Weight (g/mol): 222.31
Molecular Formula: C13H20NO2
InChi Key: TYYCBAISLMKLMT-NSHDSACASA-O
SMILES: CC(C)(C)OC(=O)C[C@H]([NH3+])C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)