accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12069-85-1

HfC

CAS RN 12069-85-1

IUPAC Name: methane hafnium
Synonyms: hafnium(IV) carbide
Molecular Weight (g/mol): 194.53
Molecular Formula: CH4Hf
InChi Key: WHJFNYXPKGDKBB-UHFFFAOYSA-N
SMILES: C.[Hf]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products