accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1207346-29-9

OSOOCl

CAS RN 1207346-29-9

IUPAC Name: oxolane-3-sulfonyl chloride
Synonyms: oxolane-3-sulfonyl chloride
Molecular Weight (g/mol): 170.61
Molecular Formula: C4H7ClO3S
InChi Key: MVZIUOJNVVIOEY-UHFFFAOYNA-N
SMILES: ClS(=O)(=O)C1CCOC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products