man

Test

CAS RN 120737-59-9

CH3NHNOOCH3H3CCH3

CAS RN 120737-59-9

IUPAC Name: tert-butyl 3-methylpiperazine-1-carboxylate
Synonyms: tert-butyl 3-methylpiperazine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: FMLPQHJYUZTHQS-UHFFFAOYNA-N
SMILES: CC1CN(CCN1)C(=O)OC(C)(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results