accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 120985-68-4

CH3FFFOOH

CAS RN 120985-68-4

IUPAC Name: 5-methyl-2-(trifluoromethyl)benzoic acid
Synonyms: 5-methyl-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: YYEMDMKMHBXYPV-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products