accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 121034-89-7

H3CNCH3H3CHNHBrBr

CAS RN 121034-89-7

IUPAC Name: (5-{[(adamantan-1-yl)methyl]amino}pentyl)trimethylazanium hydrobromide bromide
Synonyms: {5-[(adamantan-1-ylmethyl)amino]pentyl}trimethylazanium hydrobromide bromide
Molecular Weight (g/mol): 454.34
Molecular Formula: C19H38Br2N2
InChi Key: CQTDZUSQSTUZDA-UHFFFAOYSA-M
SMILES: Br.[Br-].C[N+](C)(C)CCCCCNCC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products