accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1211530-89-0

NOOHFBr

CAS RN 1211530-89-0

IUPAC Name: 2-bromo-3-fluoropyridine-4-carboxylic acid
Synonyms: 2-bromo-3-fluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: DHFIVEUMGCJILQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Br)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products