CAS RN 1212136-00-9
CAS RN 1212136-00-9
IUPAC Name:
methyl 3-[bis(4-methoxyphenyl)methyl]-2,2-dioxo-1,2λ⁶,3-oxathiazolidine-4-carboxylate
Synonyms:
methyl 3-[bis(4-methoxyphenyl)methyl]-2,2-dioxo-1,2λ⁶,3-oxathiazolidine-4-carboxylate
Molecular Weight (g/mol):
407.44
Molecular Formula:
C19H21NO7S
InChi Key:
RTHSATBSXDKZBN-UHFFFAOYNA-N
SMILES:
COC(=O)C1COS(=O)(=O)N1C(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals
CAS: 1212136-00-9 Molecular Formula: C19H21NO7S Molecular Weight (g/mol): 407.437 MDL Number: MFCD09864994 InChI Key: RTHSATBSXDKZBN-QGZVFWFLSA-N Synonym: methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate,methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate PubChem CID: 46172874 IUPAC Name: methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate SMILES: COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC