accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 121247-01-6

FOO

CAS RN 121247-01-6

IUPAC Name: 2-(3-fluorophenyl)-2-oxoacetaldehyde hydrate
Synonyms: 2-(3-fluorophenyl)-2-oxoacetaldehyde hydrate
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: VGFHAWFFIQFSRR-UHFFFAOYSA-N
SMILES: O.FC1=CC=CC(=C1)C(=O)C=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products