CAS RN 1214330-90-1
CAS RN 1214330-90-1
IUPAC Name:
1-(2-bromo-6-fluorophenyl)methanamine hydrochloride
Synonyms:
1-(2-bromo-6-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol):
240.5
Molecular Formula:
C7H8BrClFN
InChi Key:
WEGQVRTUTQKDNY-UHFFFAOYSA-N
SMILES:
Cl.NCC1=C(F)C=CC=C1Br
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Bromo-6-fluorobenzylamine hydrochloride, 96%
CAS: 1214330-90-1 Molecular Formula: C7H8BrClFN Molecular Weight (g/mol): 240.5 InChI Key: WEGQVRTUTQKDNY-UHFFFAOYSA-N Synonym: 2-bromo-6-fluorobenzylamine hcl,2-bromo-6-fluorophenyl methanamine hydrochloride,2-bromo-6-fluorophenyl methanamine;hydrochloride,1-2-bromo-6-fluorophenyl methanamine hydrochloride PubChem CID: 118798502 IUPAC Name: (2-bromo-6-fluorophenyl)methanamine;hydrochloride SMILES: C1=CC(=C(C(=C1)Br)CN)F.Cl