man

Test

CAS RN 121933-60-6

H3CNOS

CAS RN 121933-60-6

IUPAC Name: 4-methyl-4H-thieno[3,2-b]pyrrole-5-carbaldehyde
Synonyms: 4-methylthieno[3,2-b]pyrrole-5-carbaldehyde
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: XFKPNQZVRPOJNA-UHFFFAOYSA-N
SMILES: CN1C(C=O)=CC2=C1C=CS2

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results