CAS RN 122-72-5
CAS RN 122-72-5
IUPAC Name:
3-phenylpropyl acetate
Synonyms:
hydrocinnamyl acetate
Molecular Weight (g/mol):
178.23
Molecular Formula:
C11H14O2
InChi Key:
JRJGKUTZNBZHNK-UHFFFAOYSA-N
SMILES:
CC(=O)OCCCC1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Phenyl-1-propyl acetate, 98%
CAS: 122-72-5 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.231 MDL Number: MFCD00026216 InChI Key: JRJGKUTZNBZHNK-UHFFFAOYSA-N Synonym: hydrocinnamyl acetate,benzenepropanol, acetate,phenylpropyl acetate,3-phenyl-1-propyl acetate,3-acetoxypropyl benzene,3-phenyl-1-propanol, acetate,3-acetoxy-1-phenylpropane,1-acetoxy-3-phenylpropane,benzenepropanol, 1-acetate,benzenepropyl acetate PubChem CID: 31226 ChEBI: CHEBI:81257 IUPAC Name: 3-phenylpropyl acetate SMILES: CC(=O)OCCCC1=CC=CC=C1