accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 122049-54-1

OOO

CAS RN 122049-54-1

IUPAC Name: 2-oxo-3-phenylpropanoate
Synonyms: β-phenylpyruvate calcium salt phenylpyruvic acid phenylpyroracemic acid phenylpyroracemate phenylpyruvate (substance) keto-phenylpyruvate sodium salt phenylpyruvic acid phenylpyruvic acid (substance)
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H7O3
InChi Key: BTNMPGBKDVTSJY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(=O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products