accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 122138-83-4

ONHNOOF

CAS RN 122138-83-4

IUPAC Name: 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 198.15
Molecular Formula: C8H7FN2O3
InChi Key: PEOVRVSERLAIQM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C2OCCNC2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products