accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1225069-85-1

BOOH3CCH3H3CH3CNHOFFF

CAS RN 1225069-85-1

IUPAC Name: N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-3-(trifluoromethyl)benzamide
Synonyms: N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 391.2
Molecular Formula: C20H21BF3NO3
InChi Key: IUZSQEGKYZLEDM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(NC(=O)C2=CC(=CC=C2)C(F)(F)F)=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products