man

Test

CAS RN 12263-33-1

PPMnMnMn

CAS RN 12263-33-1

IUPAC Name: trimanganese(2+) diphosphanetriide
Synonyms: trimanganese(2+) diphosphanetriide
Molecular Weight (g/mol): 226.76
Molecular Formula: Mn3P2
InChi Key: ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
SMILES: [P-3].[P-3].[Mn++].[Mn++].[Mn++]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results