CAS RN 122852-69-1
CAS RN 122852-69-1
IUPAC Name:
5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-1-one hydrochloride
Synonyms:
alosetron hydrochloride
Molecular Weight (g/mol):
330.82
Molecular Formula:
C17H19ClN4O
InChi Key:
FNYQZOVOVDSGJH-UHFFFAOYSA-N
SMILES:
Cl.CN1C2=C(C3=CC=CC=C13)C(=O)N(CC1=C(C)NC=N1)CC2
1
–
3
of
3
results
1
Thermo Scientific Chemicals Alosetron hydrochloride
CAS: 122852-69-1 Molecular Formula: C17H19ClN4O
2
Alosetron Hydrochloride, Mikromolâ„¢
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Medchem Express Alosetron Hydrochloride, MedChemExpress
MedChemExpress Alosetron Hydrochloride (GR 68755C) is a potent and highly selective serotonin 5-HT3 receptor antagonist.
