man

Test

CAS RN 123-00-2

H2NNO

CAS RN 123-00-2

IUPAC Name: 3-(morpholin-4-yl)propan-1-amine
Synonyms: 4-morpholinepropanamine
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: UIKUBYKUYUSRSM-UHFFFAOYSA-N
SMILES: NCCCN1CCOCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results