man

Test

CAS RN 123-83-1

H3CH2NNHCH3CH3

CAS RN 123-83-1

IUPAC Name: [2-(dimethylazaniumyl)ethyl](ethyl)azanium
Synonyms: [2-(dimethylammonio)ethyl](ethyl)azanium
Molecular Weight (g/mol): 118.22
Molecular Formula: C6H18N2
InChi Key: WLNSKTSWPYTNLY-UHFFFAOYSA-P
SMILES: CC[NH2+]CC[NH+](C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results