CAS RN 1231709-21-9
CAS RN 1231709-21-9
IUPAC Name:
4-methylbenzene-1-sulfonic acid; ethyl 2-aminopent-4-enoate
Synonyms:
ethyl 2-aminopent-4-enoate; para-toluene sulfonate
Molecular Weight (g/mol):
315.38
Molecular Formula:
C14H21NO5S
InChi Key:
MEQLYDZSCPUCRY-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(N)CC=C.CC1=CC=C(C=C1)S(O)(=O)=O