CAS RN 123333-70-0
CAS RN 123333-70-0
IUPAC Name:
sodium 4-aminobenzene-1-sulfonate hydrate
Synonyms:
sodium 4-aminobenzenesulfonate hydrate
Molecular Weight (g/mol):
213.18
Molecular Formula:
C6H8NNaO4S
InChi Key:
VDGKZGAQOPUDQL-UHFFFAOYSA-M
SMILES:
O.[Na+].NC1=CC=C(C=C1)S([O-])(=O)=O