accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 123333-85-7

LiNS

CAS RN 123333-85-7

IUPAC Name: lithium(1+) cyanosulfanide hydrate
Synonyms: lithium(1+) hydrate thiocyanate
Molecular Weight (g/mol): 83.03
Molecular Formula: CH2LiNOS
InChi Key: UNTVNJAFDYEXLD-UHFFFAOYSA-M
SMILES: [Li+].O.[S-]C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products