man

Test

CAS RN 123691-14-5

NH2PHOO

CAS RN 123691-14-5

IUPAC Name: (3-aminopropyl)(cyclohexylmethyl)phosphinic acid
Synonyms: 3-aminopropyl(cyclohexylmethyl)phosphinic acid
Molecular Weight (g/mol): 219.27
Molecular Formula: C10H22NO2P
InChi Key: XOESDNIUAWGCLU-UHFFFAOYNA-N
SMILES: NCCCP(O)(=O)CC1CCCCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results