accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12392-64-2

NaNaCrONNOSOOOONNOSOOO(E/Z)(E/Z)

CAS RN 12392-64-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 880.73
Molecular Formula: C40H22CrN4Na2O10S2
InChi Key: LCZPIYCNOWJWPQ-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Cr+3].[O-]C1=CC=C2C=CC=CC2=C1N=NC1=C2C=CC=CC2=C(C=C1[O-])S([O-])(=O)=O.[O-]C1=CC=C2C=CC=CC2=C1N=NC1=C2C=CC=CC2=C(C=C1[O-])S([O-])(=O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products