CAS RN 124167-46-0
CAS RN 124167-46-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
4-Methylumbelliferyl 2-Acetamido-2-deoxy-3,4,6-tri-O-acetyl-beta-D-Galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.