CAS RN 124344-98-5
CAS RN 124344-98-5
IUPAC Name:
(1-methyl-5-phenyl-1H-pyrazol-3-yl)methanol
Synonyms:
(1-methyl-5-phenylpyrazol-3-yl)methanol
Molecular Weight (g/mol):
188.23
Molecular Formula:
C11H12N2O
InChi Key:
ZMDMCKKOZJKHKG-UHFFFAOYSA-N
SMILES:
CN1N=C(CO)C=C1C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Maybridge (1-methyl-5-phenyl-1h-pyrazol-3-yl)methanol, 97%, Thermo Scientific™
CAS: 124344-98-5 Molecular Formula: C11H12N2O Molecular Weight (g/mol): 188.23 MDL Number: MFCD08271936 InChI Key: ZMDMCKKOZJKHKG-UHFFFAOYSA-N Synonym: 1-methyl-5-phenyl-1h-pyrazol-3-yl methanol,1-methyl-5-phenylpyrazol-3-yl methanol,1h-pyrazole-3-methanol,1-methyl-5-phenyl,3-hydroxymethyl-1-methyl-5-phenylpyrazole,acmc-20mqzk,1-methyl-5-phenylpyrazol-3-yl methan-1-ol PubChem CID: 14354447 IUPAC Name: (1-methyl-5-phenylpyrazol-3-yl)methanol SMILES: CN1C(=CC(=N1)CO)C2=CC=CC=C2