accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1256152-35-8

H3CNCH3NHNNONNHNOH2CClOCH3OCH3Cl

CAS RN 1256152-35-8

IUPAC Name: N-(3-{[3-(2,6-dichloro-3,5-dimethoxyphenyl)-7-{[4-(diethylamino)butyl]amino}-2-oxo-1H,2H,3H,4H-pyrimido[4,5-d][1,3]diazin-1-yl]methyl}phenyl)prop-2-enamide
Synonyms: N-(3-{[3-(2,6-dichloro-3,5-dimethoxyphenyl)-7-{[4-(diethylamino)butyl]amino}-2-oxo-4H-pyrimido[4,5-d][1,3]diazin-1-yl]methyl}phenyl)prop-2-enamide
Molecular Weight (g/mol): 656.61
Molecular Formula: C32H39Cl2N7O4
InChi Key: DNVFTXQYIYFQBW-UHFFFAOYSA-N
SMILES: CCN(CC)CCCCNC1=NC=C2CN(C(=O)N(CC3=CC(NC(=O)C=C)=CC=C3)C2=N1)C1=C(Cl)C(OC)=CC(OC)=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products