CAS RN 1257044-40-8
CAS RN 1257044-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris™ ABT 199
Selective, high affinity Bcl-2 inhibitor; orally bioavailable
2
Medchem Express Venetoclax, MedChemExpress
MedChemExpress Venetoclax (ABT-199; GDC-0199) is a highly potent, selective and orally bioavailable Bcl-2 inhibitor with a Ki of less than 0.01 nM. Venetoclax induces autophagy.
