CAS RN 1257256-44-2
CAS RN 1257256-44-2
IUPAC Name:
N-(3-chloro-4-methylphenyl)-5-(4-nitrophenyl)-1,3,4-oxadiazol-2-amine
Synonyms:
N-(3-chloro-4-methylphenyl)-5-(4-nitrophenyl)-1,3,4-oxadiazol-2-amine
Molecular Weight (g/mol):
330.73
Molecular Formula:
C15H11ClN4O3
InChi Key:
JYCNWQGNEJYDQS-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C=C(NC2=NN=C(O2)C2=CC=C(C=C2)[N+]([O-])=O)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ TC-G 24

CAS: 1257256-44-2 Molecular Formula: C15H11ClN4O3 Molecular Weight (g/mol): 330.728