man

Test

CAS RN 1260763-01-6

CH3NNBrN

CAS RN 1260763-01-6

IUPAC Name: 2-bromo-6-(4-methylpiperazin-1-yl)benzonitrile
Synonyms: 2-bromo-6-(4-methylpiperazin-1-yl)benzonitrile
Molecular Weight (g/mol): 280.17
Molecular Formula: C12H14BrN3
InChi Key: XDFWUVBVNHLPIB-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C(C#N)C(Br)=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results